Skip to Content
Merck
All Photos(1)

Key Documents

14419

Sigma-Aldrich

Zinc carbonate basic

technical, ≥53% Zn basis

Synonym(s):

Zinc hydroxide carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[ZnCO3]2 · [Zn(OH)2]3
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

technical

concentration

≥53% Zn

anion traces

chloride (Cl-): ≤3000 mg/kg
sulfate (SO42-): ≤1.5%

SMILES string

[Zn++].[Zn++].O[Zn]O.O[Zn]O.O[Zn]O.[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/2CH2O3.6H2O.5Zn/c2*2-1(3)4;;;;;;;;;;;/h2*(H2,2,3,4);6*1H2;;;;;/q;;;;;;;;5*+2/p-10

InChI key

UOURRHZRLGCVDA-UHFFFAOYSA-D

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service