Skip to Content
Merck
All Photos(1)

Documents

14249

Supelco

2-Furyl methyl ketone

analytical standard

Synonym(s):

2-Acetylfuran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6O2
CAS Number:
Molecular Weight:
110.11
Beilstein:
107909
EC Number:
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.506-1.508
n20/D 1.5070 (lit.)

bp

67 °C/10 mmHg (lit.)

mp

26-28 °C (lit.)

density

1.098 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CC(=O)c1ccco1

InChI

1S/C6H6O2/c1-5(7)6-3-2-4-8-6/h2-4H,1H3

InChI key

IEMMBWWQXVXBEU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

159.8 °F - closed cup

Flash Point(C)

71 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosa Perestrelo et al.
Journal of chromatography. A, 1381, 54-63 (2015-01-27)
An improved, reliable and powerful analytical strategy based on digitally controlled microextraction by packed sorbent (MEPS) combined with ultrahigh pressure liquid chromatography (UHPLC) was validated for the simultaneous identification and quantification of major furanic derivatives, namely 5-hydroxymethyl-2-furaldehyde (5HMF), 5-methyl-2-furaldehyde (5MF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service