Skip to Content
Merck
All Photos(1)

Key Documents

13451

Sigma-Aldrich

Sodium nitroprusside dihydrate

98%

Synonym(s):

Nitroprusside sodium, SNP, Sodium nitroferricyanide, Sodium pentacyanonitrosylferrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2[Fe(CN)5NO] · 2H2O
CAS Number:
Molecular Weight:
297.95
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

98%

SMILES string

O.O.[Na+].[Na+].O=N[Fe--](C#N)(C#N)(C#N)(C#N)C#N

InChI

1S/5CN.Fe.NO.2Na.2H2O/c5*1-2;;1-2;;;;/h;;;;;;;;;2*1H2/q;;;;;2*-1;2*+1;;

InChI key

OIRZWVYIQXBRFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Aamer Sandoo et al.
Arthritis research & therapy, 13(3), R99-R99 (2011-06-23)
Patients with rheumatoid arthritis (RA) are at an increased risk for cardiovascular disease (CVD). One of the earliest manifestations of CVD is endothelial dysfunction (ED). ED can occur in both the microcirculation and the macrocirculation, and these manifestations might be
Jae Hyung Kim et al.
Journal of applied physiology (Bethesda, Md. : 1985), 107(4), 1249-1257 (2009-08-08)
There is increasing evidence that upregulation of arginase contributes to impaired endothelial function in aging. In this study, we demonstrate that arginase upregulation leads to endothelial nitric oxide synthase (eNOS) uncoupling and that in vivo chronic inhibition of arginase restores
Milos R Filipovic et al.
Journal of medicinal chemistry, 56(4), 1499-1508 (2013-02-20)
Hydrogen sulfide (H2S) has been increasingly recognized as an important signaling molecule that regulates both blood pressure and neuronal activity. Attention has been drawn to its interactions with another gasotransmitter, nitric oxide (NO). Here, we provide evidence that the physiological
A Moerman et al.
British journal of anaesthesia, 111(4), 619-626 (2013-06-07)
Although both pressure and flow are considered important determinants of regional organ perfusion, the relative importance of each is less established. The aim of the present study was to evaluate the impact of variations in flow, pressure, or both on
Bas van den Bogaard et al.
American journal of hypertension, 26(4), 574-579 (2013-03-08)
Sodium nitroprusside (SNP) and labetalol are recommended for the immediate treatment of malignant hypertension. Both are intravenous agents but have different effects on systemic hemodynamics, and may have differential effects on pulse-wave reflection and pulse-pressure amplification, with consequences for peripheral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service