Skip to Content
Merck
All Photos(1)

Key Documents

13300

Sigma-Aldrich

Phenylacetonitrile

purum, ≥98.0% (GC)

Synonym(s):

Benzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2CN
CAS Number:
Molecular Weight:
117.15
Beilstein:
385941
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥98.0% (GC)

refractive index

n20/D 1.524

bp

231-233 °C (lit.)

density

1.015 g/mL at 20 °C (lit.)

SMILES string

N#CCc1ccccc1

Looking for similar products? Visit Product Comparison Guide

Other Notes

This product has been replaced by B19401-ALDRICH | Benzyl cyanide 98%
Sales restrictions may apply

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Blenn et al.
Journal of chemical ecology, 38(7), 882-892 (2012-05-17)
Egg deposition by the Large Cabbage White butterfly Pieris brassicae on Brussels sprouts plants induces indirect defense by changing the leaf surface, which arrests the egg parasitoid Trichogramma brassicae. Previous studies revealed that this indirect defense response is elicited by
Nina E Fatouros et al.
Nature, 433(7027), 704-704 (2005-02-18)
To locate their hosts, parasitic wasps can 'eavesdrop' on the intraspecific chemical communications of their insect hosts. Here we describe an example in which the information exploited by the parasitic wasp Trichogramma brassicae is a butterfly anti-aphrodisiac that is passed
Jacques Corset et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 68(5), 1340-1346 (2007-05-01)
The carbanionic species possibly formed during the first alkylation step of phenylacetonitrile lithiated anion by CH(3)I and PhCH(2)Cl in THF-hexane solution and their complexes with the lithium salt formed have been observed by infrared spectrometry and characterized by density functional
Lee Coffey et al.
Archives of microbiology, 191(10), 761-771 (2009-09-05)
Bacterial enzymes capable of nitrile hydrolysis have significant industrial potential. Microbacterium sp. AJ115, Rhodococcus erythropolis AJ270 and AJ300 were isolated from the same location in England and harbour identical nitrile hydratase/amidase gene clusters. Strain AJ270 has been well studied due
Martinus E Huigens et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(3), 820-825 (2009-01-14)
Many insects possess a sexual communication system that is vulnerable to chemical espionage by parasitic wasps. We recently discovered that a hitch-hiking (H) egg parasitoid exploits the antiaphrodisiac pheromone benzyl cyanide (BC) of the Large Cabbage White butterfly Pieris brassicae.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service