Skip to Content
Merck
All Photos(2)

Documents

12838

Millipore

Hybriscan D Kit

Cronobacter spp., suitable for food and beverages, microbiology and in situ hybriziation

Synonym(s):

Cronobacter spp. HybriScanD, Enterobacter sakazakii HybriScanD, HybriScanD Enterobacter sakazakii

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product name

HybriScanD Cronobacter spp., suitable for microbiology

technique(s)

microbe id | in situ hybriziation: suitable

Quality Level

application(s)

food and beverages
microbiology

storage temp.

2-8°C

suitability

Cronobacter spp.

General description

HybriScan D kits detect and simultaneously identify microorganisms using rRNA as the detection target. The principle is based on in situ sandwich hybridization with two specific probes. As the rRNA of dead cells is degraded in the presence of RNase, only living cells are detected. Each cell has several thousand rRNA copies, so neither PCR nor a polymerase is needed for this technology, which means that this molecular biology test is not inhibited by the sample matrix, for example food, beverages and water.

Application

Detection and identification of Cronobacter spp. (früher Enterobacter sakazakii) in food.

Features and Benefits

Sensitivity (after pre enrichment): 1 cfu/25g

Legal Information

HybriScan is a trademark of ScanBec GmbH

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Resp. Sens. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Cronobacter spp. are frequently found on vegetables, meat, fermented bread, dairy and in baby food. Consumption of contaminated powdered infant formula milk can result in sepsis, infant meningitis and necrotizing enterocolitis.

Related Content

Fast molecular screening of beverage, water, wastewater and food for microorganisms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service