Skip to Content
Merck
All Photos(2)

Key Documents

121495

Sigma-Aldrich

Rhodanile Blue

A complex of Nile Blue and Rhodamine B., Dye content 75 %

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$482.00

$482.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$482.00

About This Item

Empirical Formula (Hill Notation):
C48H48ClN5O3
Molecular Weight:
778.38
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$482.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

form

powder or crystals

composition

Dye content, 75%

λmax

552 nm
630 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].CCN(CC)c1ccc2N=C3C(Oc2c1)=CC(=N/C(=O)c4ccccc4C5=C6C=C\C(C=C6Oc7cc(ccc57)N(CC)CC)=[N+](\CC)CC)\c8ccccc38

InChI key

UYQVPCFMVWEDCA-HAIIBULWSA-M

Application

Biological stain

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manabu Kawada et al.
Anticancer research, 24(3a), 1561-1568 (2004-07-28)
Stromal cells play an important role in regulating epithelial malignancies through diffusible factors and adhesion. Modulation of the tumor-stromal cell interaction is an attractive target for new antitumor strategies. To screen for a modulator of the interaction, we have now
L Kass
Archives of pathology & laboratory medicine, 109(4), 320-324 (1985-04-01)
Using the oxazine dye rhodanile blue, large typical megakaryocytes and small megakaryocytes (micromegakaryocytes) from the bone marrows of normal persons, and from patients with a variety of preleukemic disorders, acute lymphoblastic and nonlymphoblastic leukemia, chronic granulocytic leukemia, and idiopathic thrombocytopenic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service