Skip to Content
Merck
All Photos(1)

Key Documents

12124

Supelco

N-Methyl-N-trimethylsilyltrifluoroacetamide activated III

for GC derivatization, LiChropur, activated with imidazole

Synonym(s):

N-Methyl-N-trimethylsilyltrifluoroacetamide imidazole mixture, MSTFA activated III

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Beilstein:
1941550
MDL number:
UNSPSC Code:
41115710
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.383

storage temp.

2-8°C

SMILES string

CN(C(=O)C(F)(F)F)[Si](C)(C)C

InChI

1S/C6H12F3NOSi/c1-10(12(2,3)4)5(11)6(7,8)9/h1-4H3

InChI key

MSPCIZMDDUQPGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,2,2-trifluoro-N-methyl-N-trimethylsilylacetamide (N-Methyl-N-trimethylsilyltrifluoroacetamide activated III) may be used for derivatization, in an experimental study conducted to perform determination of scopolamine in human serum, by capillary column gas chromatographic-ion trap tandem mass spectrometric.[1]

Analysis Note

Derivatizing agent for the per(trimethylsilylation) of analytes for GC and GC-MS[1]

Other Notes

Derivatizing agent for the per(trimethylsilylation) of analytes for GC and GC-MS[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.8 °F - closed cup - Solvent

Flash Point(C)

26 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stanisław Bury et al.
Physiological and biochemical zoology : PBZ, 93(2), 90-96 (2020-02-06)
Oxidative stress, the imbalance of reactive oxygen species and antioxidant capacity, may cause damage to biomolecules pivotal for cellular processes (e.g., DNA). This may impair physiological performance and, therefore, drive life-history variation and aging rate. Because aerobic metabolism is supposed
W. Schanzer et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 687, 93-93 (1996)

Articles

Results of a study involving the ability few Fluka silylating reagents to form GC-MS-compatible trimethylsilylmethyl derivatives of NSAIDs

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service