12115
Benzidine
purum p.a., ≥98.0% (N)
Synonym(s):
4,4′-Diaminobiphenyl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H12N2
CAS Number:
Molecular Weight:
184.24
Beilstein:
742770
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
grade
purum p.a.
Quality Level
Assay
≥98.0% (N)
SMILES string
Nc1ccc(cc1)-c2ccc(N)cc2
InChI
1S/C12H12N2/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8H,13-14H2
InChI key
HFACYLZERDEVSX-UHFFFAOYSA-N
Gene Information
human ... UGT1A4(54657)
Looking for similar products? Visit Product Comparison Guide
Application
Benzidine can be used to study bioactive small molecules and cell biology. Benzidine has been used to determine pollen viability and stigma receptivity of Angelica dahurica. Benzidine has also been used to study cross-linking of horseradish peroxidase adsorbed on polycationic films.
for spectrophotometric determination of CN-, NO2-, ClO3-, SO42-
Other Notes
Reagent for the det. of blood; Reagent for the det. of peroxidases.; Reagent for the spectrophotometric det. of CN-, NO2-, ClO3-, etc.
Sales restrictions may apply
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
J.L. Royer et al.
Analytical Letters, 6, 619-619 (1973)
E.A. Burns
Analytical Chemistry, 32, 1800-1800 (1960)
THE BENZIDINE TEST: A CRITICAL REVIEW.
B J CULLIFORD et al.
Journal of forensic sciences, 9(1), 175-191 (1964-01-01)
Brian J Pak et al.
PLoS neglected tropical diseases, 8(8), e3002-e3002 (2014-08-08)
Strongyloidiasis is a persistent human parasitic infection caused by the intestinal nematode, Strongyloides stercoralis. The parasite has a world-wide distribution, particularly in tropical and subtropical regions with poor sanitary conditions. Since individuals with strongyloidiasis are typically asymptomatic, the infection can
J Whysner et al.
Pharmacology & therapeutics, 71(1-2), 107-126 (1996-01-01)
The aromatic amine benzidine (BZ) has produced various tumors, including liver tumors, in mice, rats and hamsters. BZ forms DNA adducts in rodent liver, and it is positive in most genotoxicity tests. Only bladder tumors are produced in dogs and
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service