Skip to Content
Merck
All Photos(1)

Documents

11909

Supelco

Behenic acid

analytical standard

Synonym(s):

Docosanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)20COOH
CAS Number:
Molecular Weight:
340.58
Beilstein:
1792887
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

Quality Level

grade

analytical standard

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

72-80 °C (lit.)
79-82 °C

application(s)

environmental
food and beverages

format

neat

functional group

carboxylic acid

SMILES string

CCCCCCCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C22H44O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22(23)24/h2-21H2,1H3,(H,23,24)

InChI key

UKMSUNONTOPOIO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Behenic acid is a saturated fatty acid with cholesterol-raising ability in humans.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Application

Behenic acid may be used as a reference standard for the determination of behenic acid in biological samples by electron ionization-gas chromatography-mass spectrometry (EI-GC-MS), in Amazonian palm berry, Euterpe oleraceae Mart. (Acai) by reversed phase high performance liquid chromatography (RP-HPLC) with MS, and in commercial edible oilseeds by transmethylation followed by analysis using GC combined with flame ionization detector (FID).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phytochemical and nutrient composition of the freeze-dried Amazonian palm berry, Euterpe oleraceae Mart.(Acai)
Schauss AG, et al.
Journal of Agricultural and Food Chemistry, 54(22), 8598-8603 (2006)
Determination of antioxidant compounds and antioxidant activity in commercial oilseeds for food use.
Tuberoso CIG, et al.
Food Chemistry, 103(4), 1494-1501 (2007)
High sensitivity quantitative lipidomics analysis of fatty acids in biological samples by gas chromatography-mass spectrometry.
Quehenberger O, et al.
Biochimica et Biophysica Acta - Molecular and Cell Biology of Lipids, 1811(11), 648-656 (2011)
Lais Alonso et al.
International journal of pharmaceutics, 434(1-2), 391-398 (2012-06-14)
Miltefosine (MT) is an alkylphospholipid approved for breast cancer metastasis and visceral leishmaniasis treatments, although the respective action mechanisms at the molecular level remain poorly understood. In this work, the interaction of miltefosine with the lipid component of stratum corneum
Mojgan Masoodi et al.
Rapid communications in mass spectrometry : RCM, 22(2), 75-83 (2007-12-07)
Bioactive lipid mediators derived from polyunsaturated fatty acids (PUFA) exhibit a range of tissue- and cell-specific activities in many physiological and pathological processes. Electrospray ionisation tandem mass spectrometry coupled to liquid chromatography (LC/ESI-MS/MS) is a sensitive, versatile analytical methodology for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service