Skip to Content
Merck
All Photos(3)

Key Documents

114189

Sigma-Aldrich

Acetyl chloride

reagent grade, 98%

Synonym(s):

Acetic acid chloride, Acetic chloride, Ethanoyl chloride

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$56.70
500 G
$98.30
1 KG
$158.00

$56.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$56.70
500 G
$98.30
1 KG
$158.00

About This Item

Linear Formula:
CH3COCl
CAS Number:
Molecular Weight:
78.50
Beilstein:
605303
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39050501
PubChem Substance ID:
NACRES:
NA.21

$56.70

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

reagent grade

Quality Level

vapor density

2.7 (vs air)

vapor pressure

11.69 psi ( 20 °C)
32.33 psi ( 55 °C)

Assay

98%

form

liquid

autoignition temp.

1353 °F

expl. lim.

19 %

refractive index

n20/D 1.389 (lit.)

bp

52 °C (lit.)

mp

−112 °C (lit.)

density

1.104 g/mL at 25 °C (lit.)

functional group

acyl chloride

SMILES string

CC(Cl)=O

InChI

1S/C2H3ClO/c1-2(3)4/h1H3

InChI key

WETWJCDKMRHUPV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Acetyl chloride is used as an acylating reagent for electrophilic acetylation of alkynes, alkenes, arenes, saturated alkanes, enolates, and organometallics.[1]

Acetyl chloride is a colorless acyl halide. On reaction with ammonia and alcohols, it affords acetamide and esters of acetic acid, respectively.[2] Structural parameters and rotational constants of its isotopic forms have been evaluated from the microwave spectral investigations.[3]

Application

Acetyl chloride has been used in the Lepage and Roy fatty acids analysis, and robotic fatty acid analysis.[4] It may be used for the qualitative in situ synthesis of hydrogen chloride.[5]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microwave spectrum of acetyl chloride.
Sinnott, KM.
J. Chem. Phys. , 34(3), 851-861 (1961)
Acetyl Chloride-Methanol as a Convenient Reagent for: A) Quantitative Formation of Amine Hydrochlorides B) Carboxylate Ester Formation C) Mild Removal of Nt-Boc-Protective Group.
Nudelman A, et al.
Synthetic Communications, 28(3), 471-474 (1998)
Yu Hong Lin et al.
Lipids, 47(5), 527-539 (2012-03-21)
Large population studies show that polyunsaturated fatty acids are important for human health, but determining relationships between the health benefits and the fatty acid content has been hampered by the unavailability of labor-effective high-throughput technologies. An automated high throughput fatty
Acetyl Chloride
Pearlman BA
eEROS (Encyclopedia of Reagents for Organic Synthesis) (2001)
Eagleson M.
Concise Encyclopedia Chemistry, 4-4 (1994)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service