Skip to Content
Merck
All Photos(1)

Key Documents

10005

Sigma-Aldrich

Mercury(II) amidochloride

≥95.0%, powder

Synonym(s):

Amidomercury(II) chloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
$510.00

$510.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Select a Size

Change View
500 G
$510.00

About This Item

Linear Formula:
(HgNH2)Cl
CAS Number:
Molecular Weight:
252.07
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$510.00


Estimated to ship onMarch 20, 2025


Request a Bulk Order

Quality Level

Assay

≥95.0%

form

powder

reaction suitability

core: mercury
reagent type: catalyst

SMILES string

N[Hg]Cl

InChI

1S/ClH.Hg.H2N/h1H;;1H2/q;+2;-1/p-1

InChI key

WRWRKDRWMURIBI-UHFFFAOYSA-M

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Topical mercurials.
D Dyall-Smith et al.
Journal of the American Academy of Dermatology, 25(6 Pt 1), 1097-1098 (1991-12-01)
James E Melnyk et al.
Bioorganic & medicinal chemistry, 28(20), 115712-115712 (2020-10-18)
Alternative splicing of the androgen receptor (AR) is frequently observed in castration resistant prostate cancer (CRPC). One AR isoform, the AR-V7 splice variant, is a constitutively active transcription factor which lacks a ligand binding domain and is therefore undruggable. AR-V7
L F Li
Contact dermatitis, 33(6), 392-395 (1995-12-01)
Putative allergens in, clinical manifestations of, and patch tests with traditional Chinese medicinal materials (CMM) causing contact dermatitis (CD) were studied. 14 patients with CMM CD and 351 controls were patch tested with the standard series of allergens of Beijing
B K Pang et al.
Contact dermatitis, 33(6), 423-427 (1995-12-01)
Oral lichenoid lesions (OLL) or lichen-planus-like lesions are often idiopathic. Our aim was to determine whether OLL can be caused by allergy to mercury in amalgam fillings, and whether resolution of OLL occurs after replacement of amalgam with other dental
J Handley et al.
Contact dermatitis, 29(5), 258-261 (1993-11-01)
441 consecutive patients (294 female, 147 male) with suspected contact dermatitis were patch tested to the European standard series, mercury metal (1% pet), ammoniated mercury (1% pet.), and mercuric chloride (0.1% aq.). 14 patients (3.2%), 12 of whom were female

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service