Skip to Content
Merck
All Photos(1)

Key Documents

09888

Supelco

2,6-Dimethyl-3,7-octadien-2-ol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2075633
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.452

density

0.855 g/mL at 20 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C\C=C\C(C)(C)O)C=C

InChI

1S/C10H18O/c1-5-9(2)7-6-8-10(3,4)11/h5-6,8-9,11H,1,7H2,2-4H3/b8-6+

InChI key

UPUNGXVJBANUDH-SOFGYWHQSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

163.4 °F - closed cup

Flash Point(C)

73 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service