Skip to Content
Merck
All Photos(1)

Key Documents

09780

Sigma-Aldrich

Boc-12-Ado-OH

≥98.0% (TLC)

Synonym(s):

12-(Boc-amino)dodecanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$156.00

$156.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Select a Size

Change View
500 MG
$156.00

About This Item

Empirical Formula (Hill Notation):
C17H33NO4
CAS Number:
Molecular Weight:
315.45
Beilstein:
1980302
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

$156.00


Estimated to ship onMay 14, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

colorless to white

mp

78-83 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NCCCCCCCCCCCC(O)=O

InChI

1S/C17H33NO4/c1-17(2,3)22-16(21)18-14-12-10-8-6-4-5-7-9-11-13-15(19)20/h4-14H2,1-3H3,(H,18,21)(H,19,20)
1S/C17H33NO4/c1-17(2,3)22-16(21)18-14-12-10-8-6-4-5-7-9-11-13-15(19)20/h4-14H2,1-3H3,(H,18,21)(H,19,20)

InChI key

NNPOZNVKUHYVEJ-UHFFFAOYSA-N

Other Notes

Long, hydrophobic linker[1]; Synthesis of novel lipid[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Hiruma et al.
Tetrahedron, 54, 15781-15781 (1998)
I.T. Dorn et al.
Journal of the American Chemical Society, 120, 2753-2753 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service