Skip to Content
Merck
All Photos(1)

Key Documents

09659

Sigma-Aldrich

(±)-Boc-α-phosphonoglycine trimethyl ester

≥97.0% (HPLC)

Synonym(s):

(±)-Trimethyl-Boc-α-phosphonoglycinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20NO7P
CAS Number:
Molecular Weight:
297.24
Beilstein:
4296364
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

colorless to white

application(s)

peptide synthesis

SMILES string

COC(=O)C(NC(=O)OC(C)(C)C)P(=O)(OC)OC

InChI

1S/C10H20NO7P/c1-10(2,3)18-9(13)11-7(8(12)15-4)19(14,16-5)17-6/h7H,1-6H3,(H,11,13)

InChI key

LJHAPRKTPAREGO-UHFFFAOYSA-N

Other Notes

Wittig-Horner reagent for preparing (Z)-Boc-protected dehydroamino acid derivatives; [1] and saturated, unnatural amino acids by subsequent asymmetric hydrogenation. [2][3]

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K.B. Jorgensen et al.
Tetrahedron, 55, 10527-10527 (1999)
U. Schmidt et al.
Synthesis, 1025-1025 (1992)
U. Schmidt et al.
Synthesis, 487-487 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service