Skip to Content
Merck
All Photos(1)

Key Documents

08354

Sigma-Aldrich

Methyltrioctylammonium thiosalicylate

≥95% (C)

Synonym(s):

Methyltrioctylammonium 2-mercaptobenzoate, Trioctylmethylammonium thiosalicylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H59NO2S
CAS Number:
Molecular Weight:
521.88
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (C)

impurities

≤2% water

mp

<10 °C

functional group

amine

SMILES string

[O-]C(=O)c1ccccc1S.CCCCCCCC[N+](C)(CCCCCCCC)CCCCCCCC

InChI

1S/C25H54N.C7H6O2S/c1-5-8-11-14-17-20-23-26(4,24-21-18-15-12-9-6-2)25-22-19-16-13-10-7-3;8-7(9)5-3-1-2-4-6(5)10/h5-25H2,1-4H3;1-4,10H,(H,8,9)/q+1;/p-1

InChI key

NJOSLBYBLJVAMY-UHFFFAOYSA-M

Other Notes

Ionic Liquid which can extract heavy metals (Pb, Cu, Cd)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Functionalized imidazolium cations with thioether, urea, or thiourea derivatized side chains act as metal-ligating moieties, whereas the PF6– anions provide the desired water immiscibility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service