Skip to Content
Merck
All Photos(1)

Key Documents

08268

Supelco

Cyclopentanol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9OH
CAS Number:
Molecular Weight:
86.13
Beilstein:
1900556
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.453 (lit.)
n20/D 1.453

bp

139-140 °C (lit.)

mp

−19 °C (lit.)

density

0.948 g/mL at 20 °C
0.949 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

OC1CCCC1

InChI

1S/C5H10O/c6-5-3-1-2-4-5/h5-6H,1-4H2

InChI key

XCIXKGXIYUWCLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopentanol is a cyclic alcohol utilized as an essential intermediate in the production of dyes, perfumes, and drugs. It is an important chemical used as an industrial solvent.

Application

Cyclopentanol may be used as an analytical reference standard for the determination of the analyte in frozen and dried scallops (Patinopecten yessoensis) by gas chromatography/mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermodynamic analysis of synthesis of cyclopentanol from cyclopentene and comparison with experimental data
Yao B, et al.
Applied Petrochemical Research, 5(2), 135-142 (2015)
Soft or hard ionization of molecules in helium nanodroplets? An electron impact investigation of alcohols and ethers
Yang S, et al.
Physical Chemistry Chemical Physics, 7(24), 4082-4088 (2005)
Analysis of volatile components in frozen and dried scallops (Patinopecten yessoensis) by gas chromatography/mass spectrometry
Chung HY, et al.
Food Research International, 35(1), 43-53 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service