Skip to Content
Merck

Skip To

08138

Bisphenol BP

analytical standard

Synonym(s):

1,1-Bis(4-hydroxyphenyl)-1,1-diphenylmethane, 4,4′-(Diphenylmethylene)diphenol, 4,4′-Dihydroxytetraphenylmethane, Bis(4-hydroxyphenyl)diphenylmethane, NSC 30848

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 mg
Check Cart for Availability
$198.00

About This Item

Empirical Formula (Hill Notation):
C25H20O2
CAS Number:
Molecular Weight:
352.43
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2059947

$198.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OC(C=C1)=CC=C1C(C2=CC=C(O)C=C2)(C3=CC=CC=C3)C4=CC=CC=C4

InChI

1S/C25H20O2/c26-23-15-11-21(12-16-23)25(19-7-3-1-4-8-19,20-9-5-2-6-10-20)22-13-17-24(27)18-14-22/h1-18,26-27H

InChI key

BATCUENAARTUKW-UHFFFAOYSA-N

General description

Bisphenol BP belongs to the class of bisphenols.

Application

Bisphenol BP may be used as a reference standard in the determination of bisphenol BP in serum samples using high performance liquid chromatography coupled with tandem mass spectrometry (HPLC-MS/MS).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0448750877797219
application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

cleaning products
cosmetics
food and beverages
personal care

application(s)

-

application(s)

-

format

neat

format

neat

format

neat

format

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

analytical standard

grade

-

assay

≥98.0% (HPLC)

assay

≥98.0% (HPLC)

assay

≥98.0% (HPLC)

assay

99%

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters