Skip to Content
Merck
All Photos(2)

Key Documents

07849

Supelco

Norquetiapine

analytical standard

Synonym(s):

11-(Piperazin-1-yl)dibenzo[b,f][1,4]thiazepine, N-Desalkylquetiapine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H17N3S
CAS Number:
Molecular Weight:
295.40
Beilstein:
1223426
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical
veterinary

format

neat

storage temp.

−20°C

SMILES string

C1(N2CCNCC2)=NC(C=CC=C3)=C3SC4=C1C=CC=C4

InChI

1S/C17H17N3S/c1-3-7-15-13(5-1)17(20-11-9-18-10-12-20)19-14-6-2-4-8-16(14)21-15/h1-8,18H,9-12H2

InChI key

JLOAJISUHPIQOX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service