Skip to Content
Merck
All Photos(2)

Key Documents

07682

Supelco

6-[Bis(2,2,2-trifluoroethyl)amino]-4-(trifluoromethyl)-2(1H)-quinolinone

analytical standard

Synonym(s):

LGD-2226

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$844.00

$844.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$844.00

About This Item

Empirical Formula (Hill Notation):
C14H9F9N2O
CAS Number:
Molecular Weight:
392.22
Beilstein:
10716241
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$844.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

187-190 °C

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

FC(F)(F)CN(CC(F)(F)F)c1ccc2NC(=O)C=C(c2c1)C(F)(F)F

InChI

1S/C14H9F9N2O/c15-12(16,17)5-25(6-13(18,19)20)7-1-2-10-8(3-7)9(14(21,22)23)4-11(26)24-10/h1-4H,5-6H2,(H,24,26)

InChI key

ULBPQWIGZUGPHU-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Proton NMR and Fluor NMR structures corresponds

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT RE 2 Oral

Target Organs

Prostate,Endocrine system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Gadaj et al.
MethodsX, 7, 100926-100926 (2020-06-18)
Selective androgen receptor modulators (SARMs) represent non-steroidal agents commonly abused in human and animal (i.e. equine, canine) sports, with potential for further misuse as growth promoting agents in livestock-based farming. As a direct response to the real and possible implications

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service