Skip to Content
Merck
All Photos(1)

Key Documents

07670

Supelco

N,N-Diethyl-p-phenylenediamine sulfate salt

for spectrophotometric det. of S2-, Cl2, ≥99.0%

Synonym(s):

4-Amino-N,N-diethylaniline sulfate salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NC6H4NH2 · H2SO4
CAS Number:
Molecular Weight:
262.33
Beilstein:
4219768
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (T)
≥99.0%

quality

for spectrophotometric det. of S2-, Cl2

technique(s)

UV/Vis spectroscopy: suitable

ign. residue

≤0.05%

mp

184-186 °C (lit.)
185-187 °C

solubility

H2O: 0.1 g/mL, clear

SMILES string

OS(O)(=O)=O.CCN(CC)c1ccc(N)cc1

InChI

1S/C10H16N2.H2O4S/c1-3-12(4-2)10-7-5-9(11)6-8-10;1-5(2,3)4/h5-8H,3-4,11H2,1-2H3;(H2,1,2,3,4)

InChI key

AYLDJQABCMPYEN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Rapid and Ultra-Sensitive Colorimetric Detection of H2O2 and Glucose: Utilizes N,N-Diethyl-p-phenylenediamine sulfate salt in a ferrous-metal organic framework enhancing peroxidase-mimicking activity. This breakthrough is pivotal for biochemical assays and diagnostics, demonstrating the versatility of DEPPD sulfate salt in medical research and health monitoring (Huang P et al., 2023).

  • Cotton Textile/Iron Oxide Nanozyme Composites with Peroxidase-like Activity: Features N,N-Diethyl-p-phenylenediamine sulfate salt in the synthesis and application of advanced nanozyme composites. This innovation opens new avenues in textile engineering and catalysis, showcasing the role of DEPPD sulfate salt in the development of functional materials with environmental and industrial applications (Safarik I et al., 2021).

Other Notes

Reagent for the spectrophotometric det. of Cl2 and S2-[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F.D. Snell
Photometric and Fluorometric Methods ofAnalysis, Non-Metals null
Masoumeh Hasani et al.
Journal of hazardous materials, 157(1), 161-169 (2008-02-15)
A multicomponent analysis method based on principal component analysis-artificial neural network models (PC-ANN) is proposed for the determination of phenolic compounds. The method relies on the oxidative coupling of phenols (phenol, 2 chlorophenol, 3-chlorophenol and 4-chlorophenol) to N,N-diethyl-p-phenylenediamine in the
Z B Ogel et al.
Applied microbiology and biotechnology, 71(6), 853-862 (2006-01-04)
Scytalidium thermophilum produces an extracellular phenol oxidase on glucose-containing medium. Certain phenolic acids, specifically gallic acid and tannic acid, induce the expression of the enzyme. Production at 45 degrees C in batch cultures is growth-associated and is enhanced in the
Xiao-jie Xu et al.
Guang pu xue yu guang pu fen xi = Guang pu, 24(10), 1235-1237 (2005-03-12)
In this paper, the absorption spectra of free chlorine in the visible region has been measured after free chlorine in the solution had reacted with N,N-diethyl-1,4-phenylenediamine (DPD). The relation between the absorption spectra and peak wavelength has been discussed. From
Jessica A Kershaw et al.
Analytical and bioanalytical chemistry, 379(4), 707-713 (2004-06-04)
The effect of Cu(II) on the determination of homocysteine via its electrochemically initiated reaction with N, N-diethyl-p-phenylenediamine is examined. The presence of copper inhibited the detection process for homocysteine owing to a complexation reaction occurring. This provided an indirect route

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service