Skip to Content
Merck
All Photos(2)

Key Documents

07538

Supelco

Echinacoside

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$212.00

$212.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$212.00

About This Item

Empirical Formula (Hill Notation):
C35H46O20
CAS Number:
Molecular Weight:
786.73
Beilstein:
4778612
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

$212.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥98% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H]2OC(=O)\C=C\c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C35H46O20/c1-14-24(42)26(44)29(47)35(51-14)55-32-30(48)34(49-9-8-16-3-6-18(38)20(40)11-16)53-22(13-50-33-28(46)27(45)25(43)21(12-36)52-33)31(32)54-23(41)7-4-15-2-5-17(37)19(39)10-15/h2-7,10-11,14,21-22,24-40,42-48H,8-9,12-13H2,1H3/b7-4+/t14-,21+,22+,24-,25+,26+,27-,28+,29+,30+,31+,32+,33+,34+,35-/m0/s1

InChI key

FSBUXLDOLNLABB-ISAKITKMSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Echinacoside (ECH) is a typical phenylethanoid glycoside. It is known to contain multiple unstable chemical bonds and high reactive functional groups which aids to pathways involving degradation and metabolism. It is isolated from the stems of Cistanches salsa. It is a crude drug mostly used as antisenium and antifatigue agents.[1]

Biochem/physiol Actions

Caffeic acid glycoside found in herbal medicine. Potential therapeutic for Parkinson′s disease.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pavel Mučaji et al.
Molecules (Basel, Switzerland), 16(10), 8198-8208 (2011-10-01)
The aim of the study was a HPLC evaluation of the lipoxygenase activity inhibiting activity of a water infusion of Ligustrum vulgare L. leaves and selected isolates from it. The antiradical activity of the water infusion was determined using DPPH
Wen-Jun He et al.
Planta medica, 75(13), 1400-1404 (2009-05-27)
The aim of this study was to identify and elucidate the vasorelaxant activity of echinacoside, a phenylethanoid glycoside isolated from the medicinal herb Cistanche tubulosa, and its possible underlying mechanism on isolated rat thoracic aortic rings pre-contracted with phenylephrine (PE
Min Zhu et al.
Journal of neurochemistry, 124(4), 571-580 (2012-11-30)
Neurotrophins exert their physiological functions mainly through Trk receptors, and the neurotrophic signaling network is critical to the survival of neurons. However, therapeutic use of neurotrophins in treating neurodegenerative diseases is hampered by a number of pharmacological challenges, and the
Rong Kuang et al.
Planta medica, 75(14), 1499-1504 (2009-06-24)
We have investigated the protective effects of echinacoside (ECH), one of the phenylethanoid glycosides, on H(2)O(2)-induced cytotoxicity in the rat pheochromocytoma cell line (PC12 cells). Our data show that application of ECH to H(2)O(2)-injured PC12 cells (HIPCs) increased cell viability
Parida Yamada et al.
Planta medica, 76(14), 1512-1518 (2010-04-01)
The immediate-type allergic reaction is involved in many allergic diseases such as asthma, allergic rhinitis, and sinusitis. In this study, we investigated the effect of acteoside extracted from CISTANCHE TUBULOSA (Schrenk) R. Wight on the basophilic cell-mediated allergic reaction. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service