Skip to Content
Merck
All Photos(1)

Documents

07429

Supelco

Diethyl succinate

analytical standard

Synonym(s):

Butanedioic acid, diethyl ester, Ethyl succinate, Succinic acid, diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCOCH2CH2COOC2H5
CAS Number:
Molecular Weight:
174.19
Beilstein:
907645
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

6 (vs air)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.42 (lit.)
n20/D 1.420

bp

218 °C (lit.)

mp

−20 °C (lit.)

density

1.047 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

SMILES string

CCOC(=O)CCC(=O)OCC

InChI

1S/C8H14O4/c1-3-11-7(9)5-6-8(10)12-4-2/h3-6H2,1-2H3

InChI key

DKMROQRQHGEIOW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethyl succinate, an aliphatic dibasic carboxylic acid ester of succinic acid, is present as a volatile aroma component in grape juices, wines and distilled beverages. It can be employed as an aging marker in the study of the thermal deterioration of beer.

Application

Diethyl succinate may be used as an analytical reference standard for the determination of the analyte in wine and beer samples by various chromatography techniques.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esters of aliphatic di- and tribasic carboxylic acids
Aroma of Beer, Wine and Distilled Alcoholic Beverages, 3(1-2), 151-157 (1983)
Evaluation of beer deterioration by gas chromatography-mass spectrometry/multivariate analysis: A rapid tool for assessing beer composition
Rodrigues J, et al.
Journal of Chromatography A, 1218(7), 990-996 (2011)
Gas chromatographic analysis of furfural and hydroxymethyl-furfural in wine
Shimizu JI and Watanabe M
Agricultural and Biological Chemistry, 43(6), 1365-1366 (1979)
Analysis of wine volatile profile by purge-and-trap-gas chromatography-mass spectrometry: Application to the analysis of red and white wines from different Spanish regions
Aznar M and Arroyo T
Journal of Chromatography A, 1165(1-2), 151-157 (2007)
Fast analysis of important wine volatile compounds: Development and validation of a new method based on gas chromatographic-flame ionisation detection analysis of dichloromethane microextracts
Ortega C, et al.
Journal of Chromatography A, 923(1-2), 205-214 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service