Skip to Content
Merck

Skip To

07366

(R)-(+)-1-Phenylethanol

≥98.5% (sum of enantiomers, GC)

Synonym(s):

(+)-Methyl phenyl carbinol, (R)-(+)-α-Methylbenzyl alcohol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Check Cart for Availability
$462.00

About This Item

Empirical Formula (Hill Notation):
C8H10O
CAS Number:
Molecular Weight:
122.16
UNSPSC Code:
12352002
NACRES:
NA.22
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
2039798

$462.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.5% (sum of enantiomers, GC)

form

liquid

optical activity

[α]/D +45±2°, c = 5% in methanol

optical purity

enantiomeric ratio: ≥97:3 (GC)

refractive index

n20/D 1.527

bp

88-89 °C/10 mmHg (lit.)

mp

9-11 °C (lit.)

density

1.012 g/mL at 20 °C (lit.)

functional group

hydroxyl, phenyl

SMILES string

C[C@@H](O)c1ccccc1

InChI

1S/C8H10O/c1-7(9)8-5-3-2-4-6-8/h2-7,9H,1H3/t7-/m1/s1

InChI key

WAPNOHKVXSQRPX-SSDOTTSWSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
05512P30802256323
optical activity

[α]/D +45±2°, c = 5% in methanol

optical activity

[α]/D −45±2°, c = 5% in methanol

optical activity

-

optical activity

[α]20/D −47°, c = 2.25 in hexane

form

liquid

form

liquid

form

powder

form

liquid

mp

9-11 °C (lit.)

mp

9-11 °C (lit.)

mp

28-32 °C (lit.)

mp

-

bp

88-89 °C/10 mmHg (lit.)

bp

88-89 °C/10 mmHg (lit.)

bp

202 °C (lit.)

bp

94-95 °C/10 mmHg (lit.)

optical purity

enantiomeric ratio: ≥97:3 (GC)

optical purity

enantiomeric ratio: ≥97:3 (GC)

optical purity

-

optical purity

ee: 99% (GLC)

functional group

hydroxyl, phenyl

functional group

hydroxyl, phenyl

functional group

-

functional group

hydroxyl, phenyl


Still not finding the right product?

Explore all of our products under (R)-(+)-1-Phenylethanol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

185.0 °F - closed cup

flash_point_c

85 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters