Skip to Content
Merck
All Photos(2)

Key Documents

07297

Supelco

Xylometazoline hydrochloride

analytical standard

Synonym(s):

2-(4-tert-Butyl-2,6-dimethylbenzyl)-2-imidazoline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H24N2 · HCl
CAS Number:
Molecular Weight:
280.84
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

CC1=CC(C(C)(C)C)=CC(C)=C1CC2=NCCN2.Cl

InChI

1S/C16H24N2.ClH/c1-11-8-13(16(3,4)5)9-12(2)14(11)10-15-17-6-7-18-15;/h8-9H,6-7,10H2,1-5H3,(H,17,18);1H

InChI key

YGWFCQYETHJKNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylometazoline hydrochloride is a derivative of imidazole, and a representative of the class of α1 agonists. It is commonly used in nasal decongestants, which works on the principle of constricting the blood vessels in the mucous membranes.

Application

Xylometazoline hydrochloride may be used as a reference standard in determining the xylometazoline hydrochloride content in samples of nasal drops using high performance liquid chromatography (HPLC).

Biochem/physiol Actions

α-adrenoceptor agonist; imidazoline binding site ligand.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance liquid chromatographic method for the assay of dexamethasone and xylometazoline in nasal drops containing methyl p-hydroxybenzoate.
Milojevic Z, et al.
Journal of Chromatography A, 949(1-2), 79-82 (2002)
Decongestant activity of a new formulation of xylometazoline nasal spray: a double-blind, randomized versus placebo and reference drugs controlled, dose-effect study.
FC and GM
Drugs Under Experimental and Clinical Research, 28(1), 27-35 (2002)
Pharmaceutical Chemistry E-Book (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service