Skip to Content
Merck
All Photos(1)

Documents

06849

Sigma-Aldrich

Cap B (1-methylimidazole 16% in THF)

for oligonucleotide synthesis, filtered through a 1μm filter

Synonym(s):

1-Methylimidazole solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2
CAS Number:
Molecular Weight:
82.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

for oligonucleotide synthesis

quality

filtered through a 1μm filter

impurities

<0.01% water

SMILES string

Cn1ccnc1

InChI

1S/C4H6N2/c1-6-3-2-5-4-6/h2-4H,1H3

InChI key

MCTWTZJPVLRJOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Methylimidazole has been used in a study to elaborate on the use of differential scanning calorimetry as a new indirect method for determination of vaporization enthalpies. 1-Methylimidazole has also been used in a study that analyzed the base content in in-service oils by Fourier Transform Infrared Spectroscopy.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sadia Ehsan et al.
Journal of laboratory automation, 17(3), 201-210 (2012-02-24)
An automated FTIR method for the determination of the base content (BC(pKa)) of oils at rates of > 120 samples/h has been developed. The method uses a 5% solution of trifluoroacetic acid in 1-propanol (TFA/P) added to heptane-diluted oil to
Sergey P Verevkin et al.
The journal of physical chemistry. B, 116(14), 4276-4285 (2012-03-23)
Differential scanning calorimetry (DSC) has been used to measure enthalpies of synthesis reactions of the 1-alkyl-3-methylimidazolium bromide [C(n)mim][Br] ionic liquids from 1-methylimidazole and n-alkyl bromides (with n = 4, 5, 6, 7, and 8). The optimal experimental conditions have been
Fumitoshi Shibahara et al.
The Journal of organic chemistry, 77(19), 8815-8820 (2012-09-15)
Synthetic methods for triarylated azoles containing three different aryl groups via one-pot sequential multiple C-H bond arylations are described. The one-pot sequential diarylation of C5-monoarylated azoles was achieved by the simple sequential addition of two different aryl iodides with a
Chih-Chin Tsou et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(47), 13358-13366 (2011-10-19)
S-nitrosation of the coordinated thiolate of dinitrosyl iron complexes (DNICs) to generate S-nitrosothiols (RSNOs) was demonstrated. Transformation of [{(NO)(2)Fe(μ-StBu)}(2)] (1-tBuS) into the {Fe(NO)(2)}(9) DNIC [(NO)(2)Fe(StBu)(MeIm)] (2-MeIm) occurs under addition of 20 equiv of 1-methylimidazole (MeIm) into a solution of 1-tBuS in
Bhaskar Sharma et al.
The journal of physical chemistry. A, 115(10), 1971-1984 (2011-02-22)
Quantum chemical [MP2(FULL)/6-311++G-(d,p)] calculations are done on the binding of hydrated Li(+), Na(+), K(+), Mg(2+), Cu(+), and Zn(2+) metal ions with biologically relevant heteroaromatics such as imidazole and methylimidazole. The computed interaction energies are found to be in good agreement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service