Skip to Content
Merck
All Photos(1)

Key Documents

06722

Millipore

E.coli Fluorogenic ID Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, E. coli ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
$64.10

$64.10


Estimated to ship onMay 24, 2025


Request more information

Select a Size

Change View
25 DISCS
$64.10

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$64.10


Estimated to ship onMay 24, 2025


Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
environmental
food and beverages
pharmaceutical

microbiology

storage temp.

2-8°C

suitability

Escherichia coli
coliforms

General description

E. coli Fluorogenic ID Membrane are substrate-carrying membranes used for rapid identification and differentiation of E. coli through characteristic fluorogenic reactions while placed on the agar surface post-incubation. The membranes contain fluorogenic substrate MUG. The principle is based on the ability of E. coli to cleave MUG substrate, giving a fluorogenic emission that can be detected at 365 nm.

Application

E. coli Fluorogenic ID Membrane is used for rapid detection & confirmation of Escherichia coli in water and food samples, based on a fluorogenic method. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Pseudomonas aeruginosa (27853) - Negative

Escherichia coli (25922) - Positive

Staphylococcus aureus (25923) - Negative

Salmonella Typhimurium (14028) - Negative

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service