Skip to Content
Merck
All Photos(1)

Key Documents

06154

Supelco

Aluminum standard for AAS

analytical standard, traceable to BAM

Synonym(s):

Aluminum chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
AlCl3
CAS Number:
Molecular Weight:
133.34
MDL number:
UNSPSC Code:
41116107
eCl@ss:
38120507
PubChem Substance ID:
E Number:
E173
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

1.0 g/L Al in hydrochloric acid

technique(s)

AAS: suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

SMILES string

Cl[Al](Cl)Cl

InChI

1S/Al.3ClH/h;3*1H/q+3;;;/p-3

InChI key

VSCWAEJMTAWNJL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Application

Aluminum standard for AAS may be used for the determination of total aluminum content and the fraction of aluminum leached to the infusions of black tea, green tea, Hibiscus sabdariffa, and Ilex paraguariensis (mate) and for the analysis of aluminum in drinking water, in foods and beverages consumed in the Spanish diet as well as in lubricating oils by electrothermal atomic absorption spectrometry (ET-AAS).

Preparation Note

prepared with AlCl3 and HCl (~0.05 M)

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aluminum content in foods and beverages consumed in the Spanish diet.
Lopez FF, et al.
Journal of Food Science, 65(2), 206-210 (2000)
Determination of total aluminum, chromium, copper, iron, manganese, and nickel and their fractions leached to the infusions of black tea, green tea, Hibiscus sabdariffa, and Ilex paraguariensis (mate) by ETA-AAS.
Wrobel K, et al.
Biological trace element research, 78(1-3), 271-280 (2000)
Determination of aluminum by electrothermal atomic absorption spectroscopy in lubricating oils emulsified in a sequential injection analysis system.
Burguera, JL et al.
Talanta, 68(2), 179-186 (2005)
Development of methodologies to determine aluminum, cadmium, chromium and lead in drinking water by ET AAS using permanent modifiers.
de Almeida Pereira L, et al.
Talanta, 64(2), 395-400 (2004)
Diagnosis and management of hyperhidrosis.
R A Benson et al.
BMJ (Clinical research ed.), 347, f6800-f6800 (2013-11-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service