Skip to Content
Merck
All Photos(1)

Key Documents

05687

Millipore

Glucose Fermentation Membrane

suitable for microbiology, white colored, sterile, diameter 70 mm

Synonym(s):

Indicator membrane, ID Membranes

Sign Into View Organizational & Contract Pricing

Select a Size

25 DISCS
$33.10

$33.10


Estimated to ship onMay 29, 2025


Request more information

Select a Size

Change View
25 DISCS
$33.10

About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

$33.10


Estimated to ship onMay 29, 2025


Request more information

membrane size

70 mm

sterility

sterile

shelf life

limited shelf life, expiry date on the label

storage condition

protect from light

technique(s)

microbe id | utilization test: suitable

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

Citrobacter spp.
Enterobacter spp.
Escherichia coli
Klebsiella spp.
Neisseria spp.
Proteus spp.
Salmonella spp.
Serratia spp.
Shigella spp.
Streptococcus spp.
bacteria

General description

Glucose Fermentation Membranes are substrate-carrying ID membranes used for rapid identification and differentiation of Glucose fermenting organisms through characteristic color reactions. The membranes contain chromogenic substrates, which serve as the basis for microbe differentiation by color. The principle is based on an acid indicator reaction in which specific glucose fermenting organisms will exhibit yellow color.

Application

Glucose Fermentation ID Membrane is used for rapid detection & confirmation of glucose fermenting organisms in water and food samples. They find their application in various sectors such as the food and dairy industry, water industry, pharmaceutical laboratory testing, cosmetic industry, environmental and sanitary testing, clinical diagnostic, etc.

Analysis Note

Organisms showing positive for glucose fermentation:Citrobacter freundii (8090) Enterobacter aerogenes (13048) Escherichia coli (25922) Klebsiella pneumoniae (13883) Proteus vulgaris (13315) Salmonella typhimurium (14028) Salmonella typhi (6539) Serratia marcescens (8100) Shigella flexneri (12022)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service