Skip to Content
Merck
All Photos(1)

Key Documents

05595

Supelco

4-Phenyl-1-cyclohexene

analytical standard

Synonym(s):

3-Cyclohexen-1-ylbenzene, 4-Phenylcyclohexene

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$166.00

$166.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$166.00

About This Item

Empirical Formula (Hill Notation):
C12H14
CAS Number:
Molecular Weight:
158.24
Beilstein:
1930089
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$166.00


Estimated to ship onApril 18, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥96.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.539-1.545

format

neat

storage temp.

2-8°C

SMILES string

C1CC(CC=C1)c2ccccc2

InChI

1S/C12H14/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-5,7-8,12H,6,9-10H2

InChI key

XWCWNUSFQVJNDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Phenyl-1-cyclohexene is an analog of vinylcyclohexene.[1][2][3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential ovotoxicity of 4-vinylcyclohexene and its analog, 4-phenylcyclohexene.
Hooser SB, et al.
Toxicology and Applied Pharmacology, 119(2), 302-305 (1993)
Dermal sensitization potential and inhalation toxicological evaluation of 4-phenylcyclohexene.
Nitschke DK, et al.
American Industrial Hygiene Association Journal, 52(5), 192-197 (1991)
4-Phenylcyclohexene: 2-week inhalation toxicity and neurotoxicity studies in Swiss-Webster mice.
Beekman MJ, et al.
Food And Chemical Toxicology, 34(9), 873-881 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service