Skip to Content
Merck
All Photos(2)

Key Documents

05594

Supelco

6-Phenyl-2-thiouracil

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N2OS
CAS Number:
Molecular Weight:
204.25
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.5% (HPLC)
≥97.5% (NT)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

Oc1cc(nc(S)n1)-c2ccccc2

InChI

1S/C10H8N2OS/c13-9-6-8(11-10(14)12-9)7-4-2-1-3-5-7/h1-6H,(H2,11,12,13,14)

InChI key

XEKNACRTWJHOCE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

X Huang et al.
Journal of molecular and cellular cardiology, 32(12), 2221-2228 (2000-12-13)
Two main troponin I genes, cardiac (cTnI) and slow skeletal (ssTnI), are expressed in the mammalian heart under the control of a developmentally regulated program. ssTnI is expressed first in embryonic and fetal heart, and is then downregulated by an
R Schilt et al.
Journal of chromatography, 489(1), 127-137 (1989-04-07)
Methods are described for the screening and confirmation of residues of the thyreostatics thiouracil, methylthiouracil and propylthiouracil in urine samples of cattle at levels down to 25 micrograms/l. After a selective preconcentration of the thiol-containing thyreostatics on a mercurated affinity
Highly potent and selective inhibition of HIV-1 replication by 6-phenylthiouracil derivatives.
M Baba et al.
Antiviral research, 17(4), 245-264 (1992-04-01)
Tomás Pérez-Ruiz et al.
Electrophoresis, 26(12), 2384-2390 (2005-05-17)
This paper reports the development of a method based on capillary electrophoresis with laser-induced fluorescence detection for the simultaneous determination of thiouracil (TU) and phenylthiouracil (PhTU) with high sensitivity (nanomolar range, i.e., attomoles detected). After derivatization with 5-iodoacetamidofluorescein, the analytes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service