Skip to Content
Merck
All Photos(1)

Key Documents

05374

Supelco

Phosmet-(dimethyl-d6)

PESTANAL®, analytical standard

Synonym(s):

Phosmet-d6

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$193.00

$193.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$193.00

About This Item

Empirical Formula (Hill Notation):
C11D6H6NO4PS2
Molecular Weight:
323.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$193.00


Estimated to ship onMay 19, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

mp

70-75 °C

application(s)

agriculture

format

neat

storage temp.

2-8°C

SMILES string

O=C1N(CSP(OC([2H])([2H])[2H])(OC([2H])([2H])[2H])=S)C(C2=CC=CC=C21)=O

InChI

1S/C11H12NO4PS2/c1-15-17(18,16-2)19-7-12-10(13)8-5-3-4-6-9(8)11(12)14/h3-6H,7H2,1-2H3/i1D3,2D3

InChI key

LMNZTLDVJIUSHT-WFGJKAKNSA-N

General description

Phosmet-(dimethyl-d6) is an isotope-labeled analog of the organophosphorus insecticide, phosmet, widely used for pest control in agriculture.[1]

Application

Isotope-labeled compounds are increasingly used in isotope dilution mass spectrometry (IDMS) for the quantitative analysis of pesticides. The main advantage of using the IDMS technique is because isotope-labeled compounds have nearly the same physical properties as their non-labeled counterpart analog, and thus show identical behavior during the workup and sample preparation process. This helps in overcoming the problems of matrix effects generally encountered in the usual LC-MS/GC-MS analysis potentially resulting in biased results. By spiking the sample before workup with its isotope labeled analog, the loss of analyte that leads to matrix effects can be determined and compensated.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Surface plasmon resonance sensor for phosmet of agricultural products at the ppt detection level
Song Y, et al.
Journal of Agricultural and Food Chemistry, 61(11), 2625-2630 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service