Skip to Content
Merck
All Photos(2)

Documents

05097

Supelco

(+)-Sodium L-ascorbate

analytical standard

Synonym(s):

E301; Vitamine C sodium salt, L(+)-Ascorbic acid sodium salt, Vitamin C sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NaO6
CAS Number:
Molecular Weight:
198.11
Beilstein:
3767246
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Quality Level

grade

analytical standard

Assay

≥98.5% (TLC)

shelf life

limited shelf life, expiry date on the label

composition

Na, 10.8-12.8%

loss

≤2.0% loss on drying

mp

220 °C (dec.) (lit.)

application(s)

cleaning products
cosmetics
detection
food and beverages
personal care

format

neat

SMILES string

[Na+].OC[C@H](O)[C@H]1OC(=O)C(O)=C1[O-]

InChI

1S/C6H8O6.Na/c7-1-2(8)5-3(9)4(10)6(11)12-5;/h2,5,7-10H,1H2;/q;+1/p-1/t2-,5+;/m0./s1

InChI key

PPASLZSBLFJQEF-RXSVEWSESA-M

Looking for similar products? Visit Product Comparison Guide

General description

(+)-Sodium L-ascorbate is a sodium salt of ascorbic acid.

Application

Sodium ascorbate has been used as a reference standard for the determination of sodium ascorbate in polyvitaminated premixes utilized for the fortification of infant nutrition products by reversed-phase high performance liquid chromatography (RP-HPLC) combined with ultra-violet (UV) detection and in pharmaceutical formulations by RP-ion pair LC method. (+)-Sodium L-ascorbate may be used as an analytical standard for the identification and quantification of (+)-sodium L-ascorbate as a ferric ion chelator (FIC) by HPLC post-column method with fluorescence detection.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of water-soluble vitamins by reversed-phase high performance liquid chromatography with ultra-violet detection: application to polyvitaminated premixes.
Heudi O, et al.
Journal of Chromatography A, 1070(1-2), 49-56 (2005)
High-performance liquid chromatography method for ferric iron chelators using a post-column reaction with Calcein Blue.
Ariga T, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 985, 48-53 (2015)
M G Gioia et al.
Journal of pharmaceutical and biomedical analysis, 48(2), 331-339 (2008-02-26)
A reversed-phase ion pair liquid chromatographic method (RP-LC) for the determination of dehydroascorbic acid (DHA) and ascorbic acid (AA) and also acetaminophen, which is combined in pharmaceuticals, is proposed and validated. AA and acetaminophen were analyzed directly, while DHA was
Marina A Kozuleva et al.
FEBS letters, 588(23), 4364-4368 (2014-10-15)
O2 reduction was investigated in photosystem I (PSI) complexes isolated from cyanobacteria Synechocystis sp. PCC 6803 wild type (WT) and menB mutant strain, which is unable to synthesize phylloquinone and contains plastoquinone at the quinone-binding site A1. PSI complexes from
Aleksander Hinek et al.
Journal of dermatological science, 75(3), 173-182 (2014-07-13)
Vitamin C (L-ascorbic acid), a known enhancer of collagen deposition, has also been identified as an inhibitor of elastogenesis. Present studies explored whether and how the L-ascorbic acid derivative (+) sodium L-ascorbate (SA) would affect production of collagen and elastic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service