Skip to Content
Merck
All Photos(1)

Key Documents

04718

Supelco

{2-[(2,6-Dichlorophenyl)amino]phenyl}methanol

pharmaceutical impurity standard

Synonym(s):

Diclofenac impurity C (PhEur), 2-[(2,6-Dichlorophenyl)amino]benzenemethanol, o-(2,6-Dichloroanilino)benzyl alcohol

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$543.00

$543.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Select a Size

Change View
50 MG
$543.00

About This Item

Empirical Formula (Hill Notation):
C13H11Cl2NO
CAS Number:
Molecular Weight:
268.14
Beilstein:
2859040
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$543.00


Estimated to ship onMay 21, 2025


Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

application(s)

clinical testing

format

neat

SMILES string

ClC1=C(NC2=CC=CC=C2CO)C(Cl)=CC=C1

InChI

1S/C13H11Cl2NO/c14-10-5-3-6-11(15)13(10)16-12-7-2-1-4-9(12)8-17/h1-7,16-17H,8H2

InChI key

HFLGAAAKRFLWLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service