Skip to Content
Merck
All Photos(1)

Key Documents

04609

Supelco

O-Succinyl-L-carnitine lithium salt

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$269.00

$269.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$269.00

About This Item

Empirical Formula (Hill Notation):
C11H19NO6 · xLi+
Molecular Weight:
261.27 (free acid basis)
UNSPSC Code:
41116107
NACRES:
NA.24

$269.00


Estimated to ship onApril 06, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥94.0% (HPLC)

optical activity

[α]/D -16.0±3.0°, c = 0.1 in H2O

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

Biochem/physiol Actions

O-Succinyl-L-carnitine (C4DC) is a secondary metabolite that is not directly involved in the normal growth, development or reproduction of an organism.arises from. O-Succinyl-L-carnitine arises from the TCA cycle intermediate succinyl-CoA in plasma and liver. It is an O-acylcarnitine having succinyl as the acyl substituent. O-Succinyl-L-carnitine is used as biomarker for inborn metabolic deseases related to succinyl-CoA synthase function.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Succinyl-CoA:acetoacetate transferase deficiency: identification of a new patient with a neonatal onset and review of the literature.
Niezen-Koning, K.E., et al.
The Journal of Pediatrics, 156, 870-873 (1997)
Cristiano Rizzo et al.
Clinica chimica acta; international journal of clinical chemistry, 429, 30-33 (2013-11-26)
Methylmalonic aciduria (MMA) is one of the most frequent organic acidurias, a class of diseases caused by enzymatic defects mainly involved in the catabolism of branched-chain amino acids. Recently, mild MMA and C4-dicarboxylyl-carnitine (C4DC-C) accumulation have been reported in patients

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service