Skip to Content
Merck
All Photos(1)

Key Documents

03805

Supelco

Oenothein B

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$1,730.00

$1,730.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$1,730.00

About This Item

Empirical Formula (Hill Notation):
C68H48O44
CAS Number:
Molecular Weight:
1569.08
UNSPSC Code:
85151701
NACRES:
NA.24

$1,730.00


Estimated to ship onApril 13, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

Assay

≥80% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

food and beverages

format

neat

storage temp.

2-8°C

InChI

1S/C68H50O44/c69-22-1-14(2-23(70)39(22)79)59(93)109-56-52(92)68(112-61(95)16-5-26(73)41(81)27(74)6-16)106-34-13-103-63(97)20-11-32(46(86)50(90)38(20)37-19(64(98)107-54(34)56)9-30(77)44(84)49(37)89)104-53-21(10-31(78)45(85)51(53)91)66(100)111-58-57(110-60(94)15-3-24(71)40(80)25(72)4-15)55-33(105-67(58)101)12-102-62(96)17-7-28(75)42(82)47(87)35(17)36-18(65(99)108-55)8-29(76)43(83)48(36)88/h1-11,33-34,52,54-58,67-92,101H,12-13H2

InChI key

SHYSVHSGJVNSLX-UHFFFAOYSA-N

General description

Oenothein B is a dimeric ellagitannin compound which is found commonly in genusOenothera.[1]

Application

Oenothein B may be used as a reference standard for the determination of oenothein B in aqueous extracts of Epilobii angustifolli herba[2] and Epilobium angustifolium L.[3] by high performance thin-layer chromatography (HPTLC)-densitometry, in plant materials by HPLC combined with diode array detector (DAD) and mass spectrometry (MS)[1] and in Eucalyptus globulus leaves by HPLC-DAD coupled to electrospray ionization (EIS) MS.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Barbara Hevesi Tóth et al.
Journal of pharmaceutical and biomedical analysis, 49(1), 26-31 (2008-11-18)
Epilobium species (Onagraceae) are commonly used herbal remedies in traditional, adjuvant therapy of benignus prostate hyperplasia (BPH), however the pharmacological and clinical standardization of commercially available Epilobii herba (willow-herb) remains difficult. Willow-herb products usually consist of mixtures from various species
A Kiss et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 13(4), 284-289 (2006-02-24)
An aqueous extract of Epilobium angustifolium and its main compound oenothein B (OeB), a dimeric macrocyclic ellagitannin, are specifically able to induce the neutral endopeptidase (NEP) in prostate cancer cells. The angiotensin-converting enzyme (ACE) is not influenced. Additionally, a weak
K Aoki et al.
Biochemical and biophysical research communications, 210(2), 329-337 (1995-05-16)
Oenothein B, a macrocircular dimeric ellagitannin, was found to be a potent and specific inhibitor of poly(ADP-ribose) glycohydrolase. Oenothein B suppressed glucocorticoid-sensitive mouse mammary tumor virus (MMTV) transcription in 34I cells. This suppression was accompanied by inhibition of glucocorticoid-induced endogeneous
K Miyamoto et al.
Japanese journal of cancer research : Gann, 84(1), 99-103 (1993-01-01)
The antitumor effect of oenothein B, a macrocyclic ellagitannin from Oenothera erythrosepala Bordas, on rodent tumors was studied. Oenothein B exhibited a strong antitumor activity against MM2 ascites tumors upon intraperitoneal administration to the mice before or after the tumor
Sebastian Granica et al.
Phytochemical analysis : PCA, 23(6), 582-587 (2012-03-08)
Oenothein B (OeB) is a dimeric macrocyclic ellagitannin occurring mainly in the genus Oenothera. In the literature no validated HPLC method for quantification of this compound has been reported. To develop and validate an efficient HPLC method for quantification of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service