Skip to Content
Merck
All Photos(1)

Key Documents

03582

Sigma-Aldrich

1-(Boc-amino)cyclohexanecarboxylic acid

≥98.0% (TLC)

Synonym(s):

Boc-homocycloleucine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHC6H10CO2H
CAS Number:
Molecular Weight:
243.30
Beilstein:
4189683
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (TLC)

form

powder with small lumps

reaction suitability

reaction type: Boc solid-phase peptide synthesis

color

white

mp

176-178 °C (dec.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)NC1(CCCCC1)C(O)=O

InChI

1S/C12H21NO4/c1-11(2,3)17-10(16)13-12(9(14)15)7-5-4-6-8-12/h4-8H2,1-3H3,(H,13,16)(H,14,15)

InChI key

URBHKVWOYIMKNO-UHFFFAOYSA-N

Other Notes

Synthesis of modified peptides

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Furet et al.
Bioorganic & medicinal chemistry letters, 10(20), 2337-2341 (2000-10-31)
Based on X-ray crystal structure information, mono charged phosphinate isosteres of phosphotyrosine have been designed and incorporated in a short inhibitory peptide sequence of the Grb2-SH2 domain. The resulting compounds, by exploiting additional interactions, inhibit binding to the Grb2-SH2 domain
M Mazzeo et al.
Journal of peptide science : an official publication of the European Peptide Society, 1(5), 330-340 (1995-09-01)
The cyclic octapeptide cyclo[-Pro1-Pro-Phe-Phe-Ac6c-Ile-ala-Val8-] [C8-Ac6c], containing the Pro1-Pro-Phe-Phe sequence, followed by a bulky helicogenic C alpha,alpha-dialkylated glycine residue Ac6c [1-aminocyclohexane-1-carboxylic acid), and a D-Ala residue at position 7 has been synthesized. This cyclic peptide is a deletion analogue of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service