Skip to Content
Merck

Skip To

03482

Ethylene

purum, ≥99.9%

Synonym(s):

Ethene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
CH2=CH2
CAS Number:
Molecular Weight:
28.05
UNSPSC Code:
12142100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
200-815-3
Beilstein/REAXYS Number:
1730731
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor density

0.97 (vs air)

Quality Segment

vapor pressure

35.04 atm ( 20 °C)

grade

purum

assay

≥99.9%

autoignition temp.

842 °F

expl. lim.

36 %

impurities

CO, trace, ≤0.2% carbohydrates

bp

−104 °C (lit.)

mp

−169 °C (lit.)

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Packaging

Cylinder with net ~0.7 kg

Other Notes

Sales restrictions may apply
Z742161/Z724160 DIN 1 Regulators OR 99112 DIN 1 Valve


Still not finding the right product?

Explore all of our products under Ethylene


signalword

Danger

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

target_organs

Central nervous system

Storage Class

2A - Gases

wgk

nwg

flash_point_f

-148.0 °F - closed cup

flash_point_c

-100 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Simona M Cristescu et al.
Annals of botany, 111(3), 347-360 (2012-12-18)
In view of ethylene's critical developmental and physiological roles the gaseous hormone remains an active research topic for plant biologists. Progress has been made to understand the ethylene biosynthesis pathway and the mechanisms of perception and action. Still numerous questions
L A C J Voesenek et al.
Plant biology (Stuttgart, Germany), 15(3), 426-435 (2013-04-12)
Flooding is a widely occurring environmental stress both for natural and cultivated plant species. The primary problems associated with flooding arise due to restricted gas diffusion underwater. This hampers gas exchange needed for the critical processes of photosynthesis and respiration.
Fabrizio Costa et al.
Plant science : an international journal of experimental plant biology, 211, 1-7 (2013-08-31)
The aroma trait in apple is a key factor for fruit quality strongly affecting the consumer appreciation, and its detection and analysis is often an extremely laborious and time consuming procedure. Molecular markers associated to this trait can to date



Global Trade Item Number

SKUGTIN
237981-25G04061838786425
237981-100G04061838138521
03482-700G04061833020654

Questions

Reviews

No rating value

Active Filters