Skip to Content
Merck
All Photos(1)

Key Documents

02579

Supelco

1,1,2,2-Tetrachloroethane

standard for GC, puriss. p.a., ≥99.5% (GC)

Synonym(s):

Acetylene tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CHCl2CHCl2
CAS Number:
Molecular Weight:
167.85
Beilstein:
969206
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

standard for GC
puriss. p.a.

vapor density

5.8 (vs air)

vapor pressure

8 mmHg ( 20 °C)

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.494 (lit.)
n20/D 1.494

bp

147 °C (lit.)

mp

−43 °C (lit.)

density

1.586 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

SMILES string

ClC(Cl)C(Cl)Cl

InChI

1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H

InChI key

QPFMBZIOSGYJDE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service