Skip to Content
Merck
All Photos(1)

Key Documents

02529

Supelco

2-Ethylhexanoic acid

analytical standard

Synonym(s):

2-Ethylcaproic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$35.00
5 ML
$196.00

$35.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$35.00
5 ML
$196.00

About This Item

Linear Formula:
CH3(CH2)3CH(C2H5)CO2H
CAS Number:
Molecular Weight:
144.21
Beilstein:
1750468
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

$35.00


Estimated to ship onMay 23, 2025


Request a Bulk Order

grade

analytical standard

Quality Level

vapor density

4.98 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)
10 mmHg ( 115 °C)

Assay

≥99.5% (GC)

autoignition temp.

699 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

1.04 %, 135 °F
8.64 %, 188 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.425 (lit.)
n20/D 1.425

bp

228 °C (lit.)

density

0.903 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCC(CC)C(O)=O

InChI

1S/C8H16O2/c1-3-5-6-7(4-2)8(9)10/h7H,3-6H2,1-2H3,(H,9,10)

InChI key

OBETXYAYXDNJHR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexanoic acid is one of the di(2-ethylhexyl)phthalate (DEHP) metabolites.[1]

Application

2-Ethylhexanoic acid has been used as a reference standard for the determination of 2-ethylhexanoic acid in human urine by gas chromatography−mass spectrometry (GC-MS) in selected ion monitoring (SIM) mode.[2] It may be used as a reference standard for the analysis of 2-ethylhexanoic acid in grape musts and skins as aroma compounds released by fast acid hydrolysis of flavor precursors by GC-MS technique[3] and in Thai soy sauce by dynamic headspace (DHS) sampling, direct solvent extraction (DSE) and vacuum simultaneous steam distillation−solvent extraction (V−SDE) combined with GC-MS.[4]

Other Notes

Ion Mobility: TWCCSN2 value of 136.0 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 02529 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

237.2 °F - closed cup

Flash Point(C)

114 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous analysis of the di (2-ethylhexyl) phthalate metabolites 2-ethylhexanoic acid, 2-ethyl-3-hydroxyhexanoic acid and 2-ethyl-3-oxohexanoic acid in urine by gas chromatography-mass spectrometry.
Wahl, et al.
Journal of Chromatography. B, Biomedical Applications, 758(2), 213-219 (2001)
Optimization and evaluation of a procedure for the gas chromatographic-mass spectrometric analysis of the aromas generated by fast acid hydrolysis of flavor precursors extracted from grapes.
Ibarz, et al.
Journal of Chromatography A, 1116(1-2), 217-229 (2006)
Comparison of determination method for volatile compounds in Thai soy sauce.
Wanakhachornkrai P and Lertsiri S
Food Chemistry, 83(4), 619-629 (2003)
Asymmetric synthesis and teratogenic activity of (R)-and (S)-2-ethylhexanoic acid, a metabolite of the plasticizer di-(2-ethylhexyl) phthalate.
Hauck R-S, et al.
Life Sciences, 46(7), 513-518 (1990)
S Elss et al.
Food additives and contaminants, 21(8), 811-814 (2004-09-17)
After simultaneous distillation-extraction (SDE) of commercial baby foods (n = 20) and fruit juices (n = 15) (among them 15 and eight products labelled 'organic', respectively) from 11 different suppliers, analyses performed by coupled capillary gas chromatography-mass spectrometry (GC-MS) revealed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service