Skip to Content
Merck
All Photos(1)

Documents

01710580

Echinacoside

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H46O20
CAS Number:
Molecular Weight:
786.73
Beilstein:
4778612
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

application(s)

food and beverages

SMILES string

C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@H](OCCc3ccc(O)c(O)c3)O[C@H](CO[C@@H]4O[C@H](CO)[C@@H](O)[C@H](O)[C@H]4O)[C@H]2OC(=O)\C=C\c5ccc(O)c(O)c5)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C35H46O20/c1-14-24(42)26(44)29(47)35(51-14)55-32-30(48)34(49-9-8-16-3-6-18(38)20(40)11-16)53-22(13-50-33-28(46)27(45)25(43)21(12-36)52-33)31(32)54-23(41)7-4-15-2-5-17(37)19(39)10-15/h2-7,10-11,14,21-22,24-40,42-48H,8-9,12-13H2,1H3/b7-4+/t14-,21+,22+,24-,25+,26+,27-,28+,29+,30+,31+,32+,33+,34+,35-/m0/s1

InChI key

FSBUXLDOLNLABB-ISAKITKMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Echinacoside (ECH) is a typical phenylethanoid glycoside. It is known to contain multiple unstable chemical bonds and high reactive functional groups which aids to pathways involving degradation and metabolism. It is isolated from the stems of Cistanches salsa. It is a crude drug mostly used as antisenium and antifatigue agents.
Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference Standard in the analysis of herbal medicinal products

Biochem/physiol Actions

Caffeic acid glycoside found in herbal medicine. Potential therapeutic for Parkinson′s disease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Wenjun He et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 34(4), 476-479 (2009-05-23)
The paper reviews the recent developments in pharmacological activities of echinacoside (ECH). ECH is a phenylethanoid glycoside isolated and purified from the stems of Cistanche salsa, a parasitic plant native to northwest China, which is used as a traditional Chinese
Echinacoside rescues the SHSY5Y neuronal cells from TNFalpha-induced apoptosis.
Deng M
European Journal of Pharmacology, 505(1-3), 11-18 (2004)
Toshio Morikawa et al.
Journal of natural medicines, 68(3), 561-566 (2014-04-22)
Acylated phenylethanoid glycosides, echinacoside (1) and acteoside (2), principal constituents in stems of Cistanche tubulosa (Orobanchaceae), inhibited the increase in postprandial blood glucose levels in starch-loaded mice at doses of 250-500 mg/kg p.o. These compounds (1 and 2) also significantly improved
Dong-Mi Shin et al.
Archives of pharmacal research, 37(6), 803-812 (2013-10-03)
Echinacea purpurea has been shown to have anti-diabetic activities; for example, it activates peroxisome proliferator-activated receptor γ (PPARγ) and increases insulin-stimulated glucose uptake. Adipogenesis has been used to study the insulin signaling pathway and to screen anti-diabetic compounds. The present
Ivan Kováč et al.
The Journal of pharmacy and pharmacology, 67(1), 117-125 (2014-09-23)
Although the exact underlying mechanisms are still unknown, Plantago lanceolata L. (PL) water extracts are frequently used to stimulate wound healing and to drain abscesses. Therefore, in this experimental study the effect of PL water extract on skin wound healing was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service