Skip to Content
Merck
All Photos(1)

Key Documents

01211

Sigma-Aldrich

Sodium hydrogen difluoride

technical, ≥98% (ex dried substance)

Synonym(s):

Sodium acid fluoride, Sodium bifluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NaHF2
CAS Number:
Molecular Weight:
61.99
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

Pricing and availability is not currently available.

grade

technical

Assay

≥98% (ex dried substance)

loss

≤10% loss on drying, 105 °C, 2 h

SMILES string

[F-].[Na+].F[H]

InChI

1S/2FH.Na/h2*1H;/q;;+1/p-1

InChI key

BFXAWOHHDUIALU-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service