Skip to Content
Merck
All Photos(1)

Key Documents

00970

Supelco

N-Myristoylglycine

analytical standard

Synonym(s):

2-(Tetradecanoylamino)acetic acid, N-(1-Oxotetradecyl)glycine, N-Tetradecanoylglycine, Myristoyl glycine, NSC 622050

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H31NO3
CAS Number:
Molecular Weight:
285.42
Beilstein:
1794967
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

application(s)

clinical testing

format

neat

storage temp.

2-8°C

InChI

1S/C16H31NO3/c1-2-3-4-5-6-7-8-9-10-11-12-13-15(18)17-14-16(19)20/h2-14H2,1H3,(H,17,18)(H,19,20)

InChI key

DYUGTPXLDJQBRB-UHFFFAOYSA-N

Biochem/physiol Actions

Myristoylglycine is an acylglycine with C-14 fatty acid group as the acyl moiety. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders. They are normally produced through the action of glycine N-acyltransferase which is an enzyme that catalyzes the chemical reaction: acyl-CoA + glycine ↔ CoA + N-acylglycine.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service