Skip to Content
Merck
All Photos(1)

Key Documents

00489

Sigma-Aldrich

Ethylene

puriss., ≥99.95% (GC)

Synonym(s):

Ethene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CH2
CAS Number:
Molecular Weight:
28.05
Beilstein:
1730731
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

0.97 (vs air)

vapor pressure

35.04 atm ( 20 °C)

grade

puriss.

Assay

≥99.95% (GC)

autoignition temp.

842 °F

expl. lim.

36 %

impurities

CO2, trace
N2, trace
O2, trace
water, trace
≤0.0025% methane
≤0.042% CnHm

bp

−104 °C (lit.)

mp

−169 °C (lit.)

SMILES string

C=C

InChI

1S/C2H4/c1-2/h1-2H2

InChI key

VGGSQFUCUMXWEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

pressure tin
filling pressure at 15°C: 11 bar; content (15°C, 1 bar): 11 l

Other Notes

Sales restrictions may apply

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Gas 1 - Press. Gas Liquefied gas - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

-148.0 °F - closed cup

Flash Point(C)

-100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service