Skip to Content
Merck
All Photos(2)

Key Documents

0020-BM

Sigma-Aldrich

6-suSiaLex-C3-biot

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 MG
$1,490.00

$1,490.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
0.5 MG
$1,490.00

About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

$1,490.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

conjugate

biotin conjugate

product line

BM-Series

Assay

95% (H-NMR)

form

lyophilized powder

mol wt

1341.3 Da

manufacturer/tradename

GlycoNZ

extent of labeling

20 mol% Biotin ligand density

technique(s)

glycol-bead construction: suitable
glycol-surface construction: suitable
microarray: suitable

probe type

tetrasaccharide

probe presentation

monomeric

storage temp.

−20°C

Related Categories

General description

BM-Series: Biotinylated Monomeric Probes
These glycoconjugate probes have an elongated spacer giving them the possibility to bind avidin or streptavidin. The biotin fragment is hydrophobic enough to permit the probes to be adsorbed on SepPack C18 cartridges from aqueous solutions, and to be eluted by methanol, similarly to glycosides bearing a “Lemieux-type” space. The spacer is the normal spacer of the ligand plus the spacer of the biotin label –CO(CH2)5NH–.

Application

Glycoconjugates are used to study the functions of carbohydrates, carbohydrate-binding proteins in biological processes, and on the discovery of new carbohydrate-mediated interactions.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service