Skip to Content
Merck
All Photos(1)

Key Documents

00139

Sigma-Aldrich

7,8-Dihydro-α-ionone

technical, ≥90% (sum of isomers, GC)

Synonym(s):

4-(2,6,6-Trimethyl-2-cyclohexen-1-yl)-2-butanone

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H22O
CAS Number:
Molecular Weight:
194.31
Beilstein:
2088879
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (sum of isomers, GC)

form

liquid

refractive index

n20/D 1.477

bp

90 °C/0.1 mmHg (lit.)

density

0.922 g/mL at 20 °C (lit.)

SMILES string

CC(=O)CCC1C(C)=CCCC1(C)C

InChI

1S/C13H22O/c1-10-6-5-9-13(3,4)12(10)8-7-11(2)14/h6,12H,5,7-9H2,1-4H3

InChI key

JHJCHCSUEGPIGE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service