Skip to Content
Merck
All Photos(1)

Key Documents

40-1022

Sigma-Aldrich

Phenolphthalein solution

1 % (w/v), pH 7.8-10.0

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H14O4
CAS Number:
Molecular Weight:
318.32
Beilstein:
284423
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

form

liquid

availability

available only in Japan

concentration

1 % (w/v)

pH

7.8-10.0

SMILES string

Oc1ccc(cc1)C2(OC(=O)c3ccccc23)c4ccc(O)cc4

InChI

1S/C20H14O4/c21-15-9-5-13(6-10-15)20(14-7-11-16(22)12-8-14)18-4-2-1-3-17(18)19(23)24-20/h1-12,21-22H

InChI key

KJFMBFZCATUALV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

55.4 °F

Flash Point(C)

13 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward Bormashenko et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 10(4), 654-656 (2009-01-30)
The mechanisms of floating and sliding of liquid marbles are studied. Liquid marbles containing CaCl(2) and marbles containing NaOH water solutions float on water containing Na(2)CO(3) and an alcoholic solution of phenolphthalein with no chemical reaction. Sliding of liquid marbles
Stefano Mangani et al.
Journal of medicinal chemistry, 54(15), 5454-5467 (2011-06-24)
To identify specific bacterial thymidylate synthase (TS) inhibitors, we exploited phenolphthalein (PTH), which inhibits both bacterial and human enzymes. The X-ray crystal structure of Lactobacillus casei TS (LcTS) that binds PTH showed multiple binding modes of the inhibitor, which prevented
Wasin Wongwilai et al.
Talanta, 81(3), 1137-1141 (2010-03-20)
A modification of a webcam camera for use as a small and low cost detector was demonstrated with a simple lab-on-chip reactor. Real time continuous monitoring of the reaction zone could be done. Acid-base neutralization with phenolphthalein indicator was used
Pieter A Cohen et al.
Journal of general internal medicine, 27(1), 51-56 (2011-08-17)
Pai You Guo is a weight loss supplement manufactured in China and adulterated with the banned pharmaceutical products sibutramine and phenolphthalein. The US Food and Drug Administration (FDA) announced a voluntary recall of Pai You Guo in 2009, yet clinicians
Nantao Hu et al.
Journal of nanoscience and nanotechnology, 12(1), 99-104 (2012-04-25)
Recently, much work has focused on the exfoliation of graphene through a combination of oxidation and sonication procedures, followed by reduction through chemical methods. We demonstrated that the individual graphene oxide sheets can be readily reduced by using phenolphthalin as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service