All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C14H14N3NaO3S
CAS Number:
Molecular Weight:
327.33
Beilstein:
4732884
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:
Recommended Products
form
liquid
availability
available only in Japan
dilution
(for analytical testing)
pH
3.1-4.4
SMILES string
[Na+].CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S([O-])(=O)=O
InChI
1S/C14H15N3O3S.Na/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20;/h3-10H,1-2H3,(H,18,19,20);/q;+1/p-1/b16-15+;
InChI key
STZCRXQWRGQSJD-GEEYTBSJSA-M
Looking for similar products? Visit Product Comparison Guide
Suitability
for titration
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ya Yang et al.
Nano letters, 13(2), 803-808 (2013-01-18)
In general, methyl orange (MO) can be degraded by an electrocatalytic oxidation process driven by a power source due to the generation of superoxidative hydroxyl radical on the anode. Here, we report a hybrid energy cell that is used for
Katarzyna A Michalow et al.
Environmental science and pollution research international, 19(9), 3696-3708 (2012-10-12)
TiO(2):Nb nanopowders within a dopant concentration in the range of 0.1-15 at.% were prepared by one-step flame spray synthesis. Effect of niobium doping on structural, optical and photocatalytic properties of titanium dioxide nanopowders was studied. Morphology and structure were investigated by
Jing Cao et al.
Journal of hazardous materials, 239-240, 316-324 (2012-09-29)
Novel p-n junction photocatalysts BiOI/(BiO)2CO3 with different contents of BiOI were in situ synthesized by etching (BiO)2CO3 precursor with hydroiodic acid (HI) solution. X-ray diffraction (XRD), field emission scanning electron microscopy (FE-SEM), Fourier transform infrared spectrometry (FT-IR), energy-dispersive spectroscopy (EDS)
Tarek Alammar et al.
Nanoscale, 5(17), 8045-8055 (2013-07-23)
The phase selective synthesis of nanocrystalline TiO2, titania, in ionic liquids (ILs) is explored. The influence not only of the IL but also of the Ti-precursor, pH, and temperature is investigated. Sonochemical synthesis, microwave synthesis and conventional heating are compared.
Anjana Sarkar et al.
Nanotechnology, 23(47), 475711-475711 (2012-11-03)
The structure, composition and photocatalytic activity of TiO(2) nanoparticles annealed in various gas atmospheres (N(2), NH(3) and H(2)) were studied in this work. The effect of treatment on crystal structure, particle size, chemical composition and optical absorbance were assessed by
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service