Skip to Content
Merck
All Photos(1)

Key Documents

36-0100

Sigma-Aldrich

Yttrium(III) nitrate hexahydrate

SAJ special grade, Y(NO3)3 ≥71.0 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(NO3)3 · 6H2O
CAS Number:
Molecular Weight:
383.01
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
SAJ special grade
form:
solid
Pricing and availability is not currently available.

grade

SAJ special grade

form

solid

composition

Y(NO3)3, ≥71.0%

reaction suitability

reagent type: catalyst
core: yttrium

availability

available only in Japan

density

2.682 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

O.O.O.O.O.O.[Y+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.6H2O.Y/c3*2-1(3)4;;;;;;;/h;;;6*1H2;/q3*-1;;;;;;;+3

InChI key

QBAZWXKSCUESGU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eny Kusrini et al.
Environmental monitoring and assessment, 191(8), 488-488 (2019-07-12)
This paper presents the adsorption capacity of a biosorbent derived from the inner part of durian (Durio zibethinus) rinds, which are a low-cost and abundant agro-waste material. The durian rind sorbent has been successfully utilized to remove lanthanum (La) and
Yang Yu et al.
Journal of colloid and interface science, 530, 658-666 (2018-07-15)
Arsenic contamination due to anthropogenic and natural activities has posted a great risk to humans through several pathways. As such, development of cost-effective technologies for arsenic decontamination is very important from both social and economic standpoints. In this paper, we
Yuyang Li et al.
Contrast media & molecular imaging, 2018, 8174820-8174820 (2019-01-29)
Nanoparticles (NPs) have been used as contrast agents for several bioimaging modalities. X-ray fluorescence (XRF) tomography can provide sensitive and quantitative 3D detection of NPs. With spectrally matched NPs as contrast agents, we demonstrated earlier in a laboratory system that
Jeffrey C Everts et al.
Science advances, 7(5) (2021-02-12)
The physical behavior of anisotropic charged colloids is determined by their material dielectric anisotropy, affecting colloidal self-assembly, biological function, and even out-of-equilibrium behavior. However, little is known about anisotropic electrostatic screening, which underlies all electrostatic effective interactions in such soft
Tomoaki Harada et al.
Physical chemistry chemical physics : PCCP, 16(28), 14947-14952 (2014-06-17)
Luminescent europium (Eu) and dysprosium (Dy) doped yttrium-vanadate (Y-V) nanoparticles (NPs) were synthesized in the cavity of the protein, apoferritin. Y-V NPs were synthesized by incubating a solution of apoferritin with Y(3+) and VO3(-) ions in the presence of ethylene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service