Skip to Content
Merck
All Photos(1)

Key Documents

35-0160

Sigma-Aldrich

m-Xylene

SAJ first grade, ≥98.5%

Synonym(s):

1,3-Dimethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CH3)2
CAS Number:
Molecular Weight:
106.17
Beilstein:
605441
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
grade:
SAJ first grade
Assay:
≥98.5%
bp:
138-139 °C (lit.)
vapor pressure:
16 mmHg
Pricing and availability is not currently available.

grade

SAJ first grade

vapor density

3.7 (vs air)

vapor pressure

16 mmHg

Assay

≥98.5%

form

liquid

autoignition temp.

982 °F

expl. lim.

7 %

availability

available only in Japan

refractive index

n20/D 1.497 (lit.)

bp

138-139 °C (lit.)

mp

−48 °C (lit.)

density

0.868 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(C)c1

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3

InChI key

IVSZLXZYQVIEFR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F - closed cup

Flash Point(C)

27 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michalis Koutinas et al.
Environmental microbiology, 12(6), 1705-1718 (2010-06-18)
The structure of the extant transcriptional control network of the TOL plasmid pWW0 born by Pseudomonas putida mt-2 for biodegradation of m-xylene is far more complex than one would consider necessary from a mere engineering point of view. In order
Riccardo Salvio et al.
The Journal of organic chemistry, 76(13), 5438-5443 (2011-05-27)
Artificial phosphodiesterases that combine a guanidinium unit with a general base connected by a m-xylylene linker catalyze the transesterification of the RNA model compound 2-hydroxypropyl p-nitrophenyl phosphate (HPNP). The bifunctional catalysts presented in this work show varying extents of cooperation
Sulak Sumitsawan et al.
Environmental science & technology, 45(16), 6970-6977 (2011-07-19)
Titanium dioxide (TiO(2)) is a preferred catalyst for photocatalytic oxidation of many air pollutants. In an effort to enhance its photocatalytic activity, TiO(2) was modified by pulsed plasma treatment. In this work, TiO(2) nanoparticles, coated on a glass plate, were
Dragana Bozinovski et al.
FEMS microbiology ecology, 81(1), 134-144 (2012-03-01)
A sulfate-reducing consortium maintained for several years in the laboratory with m-xylene as sole source of carbon and energy was characterized by terminal restriction fragment length polymorphism (T-RFLP) fingerprinting of PCR-amplified 16S rRNA genes and stable isotope probing of proteins
Rafael Silva-Rocha et al.
Environmental microbiology, 13(9), 2389-2402 (2011-03-18)
The regulatory and metabolic networks that rule biodegradation of pollutants by environmental bacteria are wired to the rest of the cellular physiology through both transcriptional factors and intermediary signal molecules. In this review, we examine some formalisms for describing catalytic/regulatory

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service