Skip to Content
Merck
All Photos(1)

Key Documents

30-5480

Sigma-Aldrich

TWEEN® 80

SAJ first grade

Synonym(s):

POE (20) sorbitan monooleate, Polyethylene glycol sorbitan monooleate, Polyoxyethylenesorbitan monooleate, Polysorbate 80

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12161900

grade

SAJ first grade

vapor pressure

<1 mmHg ( 20 °C)

form

liquid

availability

available only in Japan

refractive index

n20/D 1.472 (lit.)

density

1.064 g/mL at 25 °C (lit.)

application(s)

cell analysis

SMILES string

CCCCCCCC/C=C/CCCCCCCC(=O)OCCOCC(C1C(CC(O1)OCCO)OCCO)OCCO

InChI

1S/C32H60O10/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-30(36)40-25-24-37-27-29(39-22-19-34)32-28(38-21-18-33)26-31(42-32)41-23-20-35/h9-10,28-29,31-35H,2-8,11-27H2,1H3/b10-9+

InChI key

RGPBUVUVZKQNHD-MDZDMXLPSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

563.0 °F

Flash Point(C)

295 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qun Wang et al.
Brain research, 1285, 188-195 (2009-06-17)
The group IIA secretory phospholipase A2 (sPLA(2)-IIA) has been studied extensively because of its involvement in inflammatory processes. Up-regulation of this enzyme has been shown in a number of neurodegenerative diseases including cerebral ischemia and Alzheimer's disease. PX-18 is a
Cristina Marcos-Arias et al.
BMC complementary and alternative medicine, 11, 119-119 (2011-11-29)
Candida-associated denture stomatitis is a frequent infectious disease. Treatment of this oral condition is difficult because failures and recurrences are common. The aim of this study was to test the in vitro antifungal activity of pure constituents of essentials oils.
Martin L Cross et al.
Clinical and vaccine immunology : CVI, 16(9), 1378-1380 (2009-07-03)
Cultures of Mycobacterium bovis BCG, comprising predominantly single-cell bacilli, were prepared in broth without animal-derived reagents. When formulated into a vegetable-derived lipid matrix, the vaccine was stable in vitro and was immunogenic in vivo upon feeding it to mice. This
Rokeya Tasneen et al.
Antimicrobial agents and chemotherapy, 52(10), 3664-3668 (2008-08-13)
PA-824 is in phase II clinical testing to treat tuberculosis. At a dose of 100 mg/kg of body weight, it has demonstrated bactericidal activity during the initial and continuation phases of treatment in a murine model of tuberculosis. In a
Syed A Sattar et al.
PloS one, 6(2), e17340-e17340 (2011-03-11)
Human noroviruses (HuNoV), a major cause of acute gastroenteritis worldwide, cannot be readily cultured in the lab. Therefore, a feline calicivirus (FCV) is often used as its surrogate to, among other things, test alcohol-based handrubs (ABHR). The more recent laboratory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service